Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidodicarbonimidic diamide, N-phenyl-, hydrochloride | 55-57-2 | 99.8% | 213.67 | C8H12ClN5 | 25 G
SDP

Catalog No. 5000213462
Encompass_Preferred

Phenylbiguanide hydrochloride is the hydrochloride salt of phenylbiguanide, a selective 5-HT3 receptor agonist commonly used in receptor pharmacology and in vitro pharmacological assays. It is supplied as a research-grade reagent with supporting documentation for characterization and safe handling.

  • Selective 5-HT3 receptor agonist (EC50 ≈ 3.0 μM).
  • High purity: 99.8% by analytical testing.
  • Molecular weight: 213.67 g/mol.
  • CAS number: 55-57-2.
  • Available in multiple pack sizes, including 25 g.
  • Provided with datasheet and safety data sheet for handling and storage.

Catalog No. 50-002-13462 Supplier Medchemexpress LLC Supplier No. HY101331A25G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylbiguanide hydrochloride is the hydrochloride salt of phenylbiguanide, a selective 5-HT3 receptor agonist commonly used in receptor pharmacology and in vitro pharmacological assays. It is supplied as a research-grade reagent with supporting documentation for characterization and safe handling.

  • Selective 5-HT3 receptor agonist (EC50 ≈ 3.0 μM).
  • High purity: 99.8% by analytical testing.
  • Molecular weight: 213.67 g/mol.
  • CAS number: 55-57-2.
  • Available in multiple pack sizes, including 25 g.
  • Provided with datasheet and safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.