Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidurea, NF, 26-28%, Spectrum™ Chemical
SDP

Catalog No. 18603428 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-428 500 g Poly Bottle
18-603-429 2.5 kg Poly Pail
18-603-430 12 kg Poly Pail
18-603-431 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-428 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I1007500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

I1007, 39236-46-9, C11H16N8O8

Spectrum™ Chemical Imidurea, NF is an antimicrobial preservative used in topical pharmaceutical preparations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.30
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYNA-N
IUPAC Name 3-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1-[({[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoyl}amino)methyl]urea
SMILES OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O

Specifications

CAS Max % 100%
Ignition Residue 3%
Loss on Drying 3%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 26 to 28%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.