Learn More
Medchemexpress LLC Imifoplatin (PT-112) | 1339960-28-9 | 95.0% | 485.23 g·mol⁻¹ | C6H16N2O7P2Pt | 5 MG
SDP
Catalog No. 5000263127
Encompass_Preferred
Imifoplatin is a platinum-containing phosphaplatin research compound with reported antitumor activity and the ability to induce apoptosis and immunogenic cell death. It is provided as a high-purity reagent in a small research-scale quantity and is intended for preclinical and in vitro studies. Documentation such as the safety data sheet and certificate of analysis is available with the product.
  • Platinum-containing phosphaplatin class compound.
  • Exhibits antitumor activity and can induce apoptosis.
  • Causes immunogenic cell death in experimental models.
  • High purity: 95.0%.
  • Molecular weight 485.23 g·mol⁻¹; formula C6H16N2O7P2Pt.
  • Supplied in small, research-scale quantity suitable for preclinical use.
  • Includes safety and quality documentation (SDS, COA).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-63127 Supplier Medchemexpress LLC Supplier No. HY1091465MG
Imifoplatin is a platinum-containing phosphaplatin research compound with reported antitumor activity and the ability to induce apoptosis and immunogenic cell death. It is provided as a high-purity reagent in a small research-scale quantity and is intended for preclinical and in vitro studies. Documentation such as the safety data sheet and certificate of analysis is available with the product.
  • Platinum-containing phosphaplatin class compound.
  • Exhibits antitumor activity and can induce apoptosis.
  • Causes immunogenic cell death in experimental models.
  • High purity: 95.0%.
  • Molecular weight 485.23 g·mol⁻¹; formula C6H16N2O7P2Pt.
  • Supplied in small, research-scale quantity suitable for preclinical use.
  • Includes safety and quality documentation (SDS, COA).
  • For research use only; not for human or veterinary use.