Learn More
Medchemexpress LLC Imiloxan hydrochloride | 81167-22-8 | 98.5% | 280.75 | C14H17ClN2O2 | 10 MG
SDP
Catalog No. 5000212516
Encompass_Preferred
Imiloxan hydrochloride is a potent, selective α2B-adrenoceptor antagonist used in pharmacology research to probe adrenergic receptor subtype function. It is used in both in vitro receptor assays and in vivo models to study antagonist activity and physiological responses such as mydriasis reversal.
  • Selective α2B-adrenoceptor antagonist.
  • High purity suitable for research use.
  • Molecular weight 280.75 g·mol⁻¹.
  • Available in small, precise research quantities.
  • Applicable to in vitro assays and in vivo pharmacology models.
Catalog No. 50-002-12516 Supplier Medchemexpress LLC Supplier No. HY101337A10MG
Imiloxan hydrochloride is a potent, selective α2B-adrenoceptor antagonist used in pharmacology research to probe adrenergic receptor subtype function. It is used in both in vitro receptor assays and in vivo models to study antagonist activity and physiological responses such as mydriasis reversal.
  • Selective α2B-adrenoceptor antagonist.
  • High purity suitable for research use.
  • Molecular weight 280.75 g·mol⁻¹.
  • Available in small, precise research quantities.
  • Applicable to in vitro assays and in vivo pharmacology models.