Learn More
Medchemexpress LLC Imiloxan hydrochloride | 81167-22-8 | 98.5% | 280.75 g/mol | C14H17ClN2O2 | 5 MG
SDP
Catalog No. 5000213583
Encompass_Preferred
Imiloxan hydrochloride is a potent, selective α2B-adrenoceptor antagonist used in pharmacological research to probe α2-adrenergic receptor subtype function. It is supplied as a characterized solid with defined purity and molecular weight for use in receptor-binding studies, in vitro assays, and in vivo experiments.
  • Selective α2B-adrenoceptor antagonist
  • High purity suitable for research applications
  • Supplied in small research pack sizes for lab use
  • Supported by analytical data (HNMR, LC-MS)
  • Provided with SDS, COA, and handling instructions
Catalog No. 50-002-13583 Supplier Medchemexpress LLC Supplier No. HY101337A5MG
Imiloxan hydrochloride is a potent, selective α2B-adrenoceptor antagonist used in pharmacological research to probe α2-adrenergic receptor subtype function. It is supplied as a characterized solid with defined purity and molecular weight for use in receptor-binding studies, in vitro assays, and in vivo experiments.
  • Selective α2B-adrenoceptor antagonist
  • High purity suitable for research applications
  • Supplied in small research pack sizes for lab use
  • Supported by analytical data (HNMR, LC-MS)
  • Provided with SDS, COA, and handling instructions