Learn More
Medchemexpress LLC Imiloxan hydrochloride | 81167-22-8 | 98.5% | 280.75 g/mol | C14H17ClN2O2 | 50 MG
SDP
Catalog No. 5000212533
Encompass_Preferred
Imiloxan hydrochloride is a research chemical that acts as a selective α2B-adrenoceptor antagonist, supplied as a solid and characterized for laboratory pharmacology studies. It is used to probe adrenergic receptor subtype function in vitro and in vivo, and is accompanied by analytical and safety documentation for research use only.
  • Selective α2B-adrenoceptor antagonist for receptor subtype studies.
  • High purity suitable for in vitro and in vivo experiments.
  • Available in small milligram pack sizes for flexible dosing and formulation.
  • Supplied with a product data sheet and safety data sheet for laboratory handling.
  • Solid form enables precise weighing and preparation of solutions.
Catalog No. 50-002-12533 Supplier Medchemexpress LLC Supplier No. HY101337A50MG
Imiloxan hydrochloride is a research chemical that acts as a selective α2B-adrenoceptor antagonist, supplied as a solid and characterized for laboratory pharmacology studies. It is used to probe adrenergic receptor subtype function in vitro and in vivo, and is accompanied by analytical and safety documentation for research use only.
  • Selective α2B-adrenoceptor antagonist for receptor subtype studies.
  • High purity suitable for in vitro and in vivo experiments.
  • Available in small milligram pack sizes for flexible dosing and formulation.
  • Supplied with a product data sheet and safety data sheet for laboratory handling.
  • Solid form enables precise weighing and preparation of solutions.