Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imiloxan hydrochloride | 81167-22-8 | MFCD01074788 | >98.0% | 280.75 | C14H17ClN2O2 | 25 MG
SDP

Catalog No. 5000212815
Encompass_Preferred

Imiloxan hydrochloride is the hydrochloride salt of imiloxan, a potent and selective α2B-adrenoceptor antagonist used in pharmacology research. Supplied as a solid reagent, it is intended for distinguishing α2 receptor subtypes and for probing adrenergic signaling in both in vitro and in vivo studies.

  • Potent, selective α2B-adrenoceptor antagonist for receptor pharmacology.
  • Suitable for both in vitro and in vivo pharmacodynamic studies.
  • High purity (≥98% HPLC) supporting reproducible results.
  • Hydrochloride salt form improves aqueous solubility for dosing.
  • Available in small research pack sizes appropriate for screening and dosing.
  • Molecular formula C14H17ClN2O2; molecular weight 280.75 g/mol.

Catalog No. 50-002-12815 Supplier Medchemexpress LLC Supplier No. HY101337A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imiloxan hydrochloride is the hydrochloride salt of imiloxan, a potent and selective α2B-adrenoceptor antagonist used in pharmacology research. Supplied as a solid reagent, it is intended for distinguishing α2 receptor subtypes and for probing adrenergic signaling in both in vitro and in vivo studies.

  • Potent, selective α2B-adrenoceptor antagonist for receptor pharmacology.
  • Suitable for both in vitro and in vivo pharmacodynamic studies.
  • High purity (≥98% HPLC) supporting reproducible results.
  • Hydrochloride salt form improves aqueous solubility for dosing.
  • Available in small research pack sizes appropriate for screening and dosing.
  • Molecular formula C14H17ClN2O2; molecular weight 280.75 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.