Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodiacetic acid disodium salt hydrate, 97%

Catalog No. AA4380930
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4380930 250 g
AA4380918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4380930 Supplier Thermo Scientific Chemicals Supplier No. 04380930
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17593-73-6 | C4H5NNa2O4 | 177.07 g/mol

Sodium iminodiacetate dibasic hydrate (Iminodiacetic acid disodium salt monohydrate) has been used in the preparation of different iminodiacetic acid-agarose supports, useful for selective adsorption of large proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17593-73-6
Molecular Formula C4H5NNa2O4
Molecular Weight (g/mol) 177.07
MDL Number MFCD00150689
InChI Key HAXVIVNBOQIMTE-UHFFFAOYSA-L
Synonym disodium iminodiacetate hydrate, iminodiacetic acid disodium salt hydrate, sodium 2,2'-azanediyldiacetate hydrate, disodium hydrate iminodiacetate, iminodiacetic acid, disodium salt monohydrate, sodium iminodiacetate dibasic hydrate, acmc-209ri5, c4h5no4.2na.h2o, disodium iminodiacetate monohydrate, sodium 2,2-azanediyldiacetate hydrate
PubChem CID 12520792
IUPAC Name disodium;2-(carboxylatomethylamino)acetate;hydrate
SMILES [Na+].[Na+].[O-]C(=O)CNCC([O-])=O

Specifications

Quantity 250 g
Merck Index 14,4917
Solubility Information Soluble in water.
Formula Weight 177.07 (Anhydrous)
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Iminodiacetic acid disodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.