Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodiacetic Acid 98.0+%, TCI America™
SDP

Catalog No. I000625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000625G 25 g
I0006500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000625G Supplier TCI America Supplier No. I000625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point 250°C
Color White
Quantity 25 g
Formula Weight 133.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iminodiacetic Acid
EINECSNumber (2)-1211
RTECSNumber AI2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.