Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iminodibenzyl | 494-19-9 | 100.0% | 250 G
SDP

Catalog No. 5000414222
Encompass_Preferred

Iminodibenzyl is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is used as an intermediate of the antiepileptic agent Carbamazepine. This product is for research use only.

  • Purity: 99.98%
  • Available in various quantities
  • Also available as a solution or solid with solvent
  • Solid appearance, off-white to yellow color
  • Can be dissolved in DMSO (100 mg/mL)
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-14222 Supplier Medchemexpress LLC Supplier No. 250GHYB2131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iminodibenzyl is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is used as an intermediate of the antiepileptic agent Carbamazepine. This product is for research use only.

  • Purity: 99.98%
  • Available in various quantities
  • Also available as a solution or solid with solvent
  • Solid appearance, off-white to yellow color
  • Can be dissolved in DMSO (100 mg/mL)
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.