Learn More
Medchemexpress LLC Iminodibenzyl | 494-19-9 | 99.9% | 1000 G
SDP
Catalog No. 5000410343
Encompass_Preferred
Iminodibenzyl is a biochemical reagent used as a biological material or organic compound for life science related research. It serves as an intermediate for the antiepileptic agent Carbamazepine and is intended for research use only.
  • Used as a biological material.
  • Functions as an organic compound for life science research.
  • Essential intermediate in the synthesis of antiepileptic agents.
  • High purity of 99.98%.
  • Available in solid form for various applications.
  • Specific storage recommendations ensure product stability.
Catalog No. 50-004-10343 Supplier Medchemexpress LLC Supplier No. HYB21311000G
Iminodibenzyl is a biochemical reagent used as a biological material or organic compound for life science related research. It serves as an intermediate for the antiepileptic agent Carbamazepine and is intended for research use only.
  • Used as a biological material.
  • Functions as an organic compound for life science research.
  • Essential intermediate in the synthesis of antiepileptic agents.
  • High purity of 99.98%.
  • Available in solid form for various applications.
  • Specific storage recommendations ensure product stability.