Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminostilbene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019209 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9209 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9209 Supplier MilliporeSigma Supelco Supplier No. PHR1197500MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards..

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 256-96-2
Physical Form Neat
Linear Formula C14H11 N
MDL Number MFCD00005071
Melting Point 196°C to 199°C
Synonym 5 H-Dibenz[b,f]azepine; Carbamazepine Related Compound B; Iminostilbene
Type Certified Reference Material
Quantity 500 mg
Formula Weight 193.24
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.