Learn More
Lkt Laboratories Imipenem Monohydrate, ≥98%, 25MG
SDP
Catalog No. NC9737752
Encompass_Preferred
Carbapenem β-lactam; penicillin binding protein inhibitor. CAS: 74431-23-5. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9737752 Supplier Lkt Laboratories Supplier No. I493425MG
Carbapenem β-lactam; penicillin binding protein inhibitor. CAS: 74431-23-5. Additional sizes available upon request. Ships from Saint Paul, MN.