Learn More
Selleck Chemical LLC Imipenem-5mg
SDP
Catalog No. 502389636
Encompass_Preferred
Imipenem acts as an inhibitor of AmpC β-lactamase and is an antibiotic useful for the treatment of a number of bacterial infections.
Catalog No. 50-238-9636 Supplier Selleck Chemical LLC Supplier No. S36675MG
Imipenem acts as an inhibitor of AmpC β-lactamase and is an antibiotic useful for the treatment of a number of bacterial infections.