Learn More
Apexbio Technology LLC Imipenem (hydrate), 100mg CAS# 74431-23-5
SDP
Supplier:  Apexbio Technology LLC B8674100
Encompass_Preferred
Imipenem is an intravenous -lactam antibiotic. It was the first member of carbapenem antibiotics. Carbapenems are highly resistant to -lactamases enzymes produced by many multi-resistant Gram-negative bacteria and therefore play a key role in treating infections that are not easily treated with other antibiotics. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8460