Learn More
Alkali Scientific Imipenem Monohydrate, 100 Mg
SDP
Supplier:  Alkali Scientific MIP003CP
Encompass_Preferred
Imipenem Monohydrate, 100 mg, is an antibiotic reagent used in microbiological research to study bacterial resistance patterns and efficacy of antibiotic treatments. As a broad-spectrum beta-lactam antibiotic, it is highly effective against a variety of both Gram-positive and Gram-negative bacterial pathogens. Imipenem inhibits the synthesis of bacterial cell walls, leading to cell death. This 100 mg concentration is used for more concentrated applications in laboratory tests, including minimal inhibitory concentration (MIC) assays and susceptibility testing. Researchers also utilize this reagent to examine resistance mechanisms and explore the use of imipenem in combination therapies against resistant strains.
Catalog No. 50-006-61654