Learn More
Alkali Scientific Imipenem Monohydrate, 5 Mg
SDP
Catalog No. 5000660051
Encompass_Preferred
Imipenem Monohydrate, 5 mg, is an antibiotic reagent used for microbiological research and susceptibility testing. It is a broad-spectrum beta-lactam antibiotic that is highly effective against a wide range of bacterial pathogens, including both Gram-positive and Gram-negative bacteria. Imipenem works by inhibiting bacterial cell wall synthesis, which leads to bacterial cell death. It is commonly used in microbiological studies for evaluating antibiotic resistance, performing minimal inhibitory concentration (MIC) tests, and assessing the effectiveness of combination therapies. Researchers also utilize this antibiotic in the study of antibiotic-resistant bacteria and to monitor the development of resistance in clinical settings.
Catalog No. 50-006-60051 Supplier Alkali Scientific Supplier No. MIP001CP
Imipenem Monohydrate, 5 mg, is an antibiotic reagent used for microbiological research and susceptibility testing. It is a broad-spectrum beta-lactam antibiotic that is highly effective against a wide range of bacterial pathogens, including both Gram-positive and Gram-negative bacteria. Imipenem works by inhibiting bacterial cell wall synthesis, which leads to bacterial cell death. It is commonly used in microbiological studies for evaluating antibiotic resistance, performing minimal inhibitory concentration (MIC) tests, and assessing the effectiveness of combination therapies. Researchers also utilize this antibiotic in the study of antibiotic-resistant bacteria and to monitor the development of resistance in clinical settings.