Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Imipenem Monohydrate, 5 Milligrams
SDP

Catalog No. 50997886
Encompass_Preferred

  • CAS Number: 74431-23-5
  • Molecular Formula: C12H17N3O4S • H2O
  • Molecular Weight: 317.36

Catalog No. 50-997-886 Supplier Research Products International Corp Supplier No. I261000.005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

  • CAS Number: 74431-23-5
  • Molecular Formula: C12H17N3O4S • H2O
  • Molecular Weight: 317.36

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.