Learn More
Medchemexpress LLC Imipenem monohydrate | 74431-23-5 | 99.9% | 25 MG
SDP
Catalog No. 5000414260
Encompass_Preferred
Imipenem monohydrate is a stable crystalline derivative of thienamycin. It is an antibiotic with excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria.
  • Can be used for the research of carbapenem-nonsusceptible infections.
  • Can be used for the research of P. aeruginosa biofilm infections.
Catalog No. 50-004-14260 Supplier Medchemexpress LLC Supplier No. 25MGHYB1369
Imipenem monohydrate is a stable crystalline derivative of thienamycin. It is an antibiotic with excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria.
  • Can be used for the research of carbapenem-nonsusceptible infections.
  • Can be used for the research of P. aeruginosa biofilm infections.