Learn More
Medchemexpress LLC Imipenem monohydrate | 74431-23-5 | 99.9% | 250 MG
SDP
Catalog No. 5000413951
Encompass_Preferred
Imipenem monohydrate is a stable crystalline derivative of thienamycin. This antibiotic exhibits excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria, making it suitable for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections.
  • Stable crystalline derivative
  • Exhibits excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria
  • Suitable for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections
  • Off-white to yellow solid appearance
  • Purity of 99.90%
  • Stable in powder form for 3 years when stored at -20°C (solutions are unstable, fresh preparation recommended)
Catalog No. 50-004-13951 Supplier Medchemexpress LLC Supplier No. 250MGHYB1369A
Imipenem monohydrate is a stable crystalline derivative of thienamycin. This antibiotic exhibits excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria, making it suitable for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections.
  • Stable crystalline derivative
  • Exhibits excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria
  • Suitable for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections
  • Off-white to yellow solid appearance
  • Purity of 99.90%
  • Stable in powder form for 3 years when stored at -20°C (solutions are unstable, fresh preparation recommended)