Learn More
Medchemexpress LLC Imipenem monohydrate | 74431-23-5 | MFCD09753321 | 99.5% | 10 MG
SDP
Catalog No. 5000297221
Encompass_Preferred
Imipenem monohydrate is a crystalline carbapenem antibiotic provided as a high-purity analytical standard for research use only. It is intended for reference standard work, method development, and microbiology research involving carbapenem activity. The material is supplied as a stable solid monohydrate suitable for analytical assays and quality control.
Catalog No. 50-002-97221 Supplier Medchemexpress LLC Supplier No. HYB1369R10MG
Imipenem monohydrate is a crystalline carbapenem antibiotic provided as a high-purity analytical standard for research use only. It is intended for reference standard work, method development, and microbiology research involving carbapenem activity. The material is supplied as a stable solid monohydrate suitable for analytical assays and quality control.