Learn More
Cayman Chemical Imipenem (hydrate), Purity: ≥98%, CAS: 74431-23-5, 100 mg
SDP
Catalog No. 502973225
Encompass_Preferred
An intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0.5-2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species
Catalog No. 50-297-3225 Supplier Cayman Chemical Supplier No. 16039100 MG
An intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0.5-2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species