Learn More
Cayman Chemical ImIpenmhydrate 10mg
SDP
Catalog No. 502972362
Encompass_Preferred
An intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0.5-2 UG/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species
Catalog No. 50-297-2362 Supplier Cayman Chemical Supplier No. 1603910 MG
An intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0.5-2 UG/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species