Learn More
Medchemexpress LLC Imipramine-d4 hydrochloride | 61361-33-9 | 99.95% | 5 MG
SDP
Catalog No. 5000411635
Encompass_Preferred
Imipramine-d4 (hydrochloride) is the deuterium-labeled Imipramine hydrochloride. Imipramine hydrochloride is an orally active tertiary amine tricyclic antidepressant and a Fascin1 inhibitor with antitumor activities. It also inhibits serotonin transporter with an IC50 value of 32 nM. Imipramine hydrochloride stimulates U-87MG glioma cells autophagy and induces HL-60 cell apoptosis, showing neuroprotective and immunomodulatory effects.
  • Deuterium labeled Imipramine hydrochloride
  • Orally active tertiary amine tricyclic antidepressant
  • Fascin1 inhibitor with antitumor activities
  • Inhibits serotonin transporter (IC50 value of 32 nM)
  • Stimulates U-87MG glioma cells autophagy
  • Induces HL-60 cell apoptosis
  • Shows neuroprotective and immunomodulatory effects
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-11635 Supplier Medchemexpress LLC Supplier No. HYB1490S5MG
Imipramine-d4 (hydrochloride) is the deuterium-labeled Imipramine hydrochloride. Imipramine hydrochloride is an orally active tertiary amine tricyclic antidepressant and a Fascin1 inhibitor with antitumor activities. It also inhibits serotonin transporter with an IC50 value of 32 nM. Imipramine hydrochloride stimulates U-87MG glioma cells autophagy and induces HL-60 cell apoptosis, showing neuroprotective and immunomodulatory effects.
  • Deuterium labeled Imipramine hydrochloride
  • Orally active tertiary amine tricyclic antidepressant
  • Fascin1 inhibitor with antitumor activities
  • Inhibits serotonin transporter (IC50 value of 32 nM)
  • Stimulates U-87MG glioma cells autophagy
  • Induces HL-60 cell apoptosis
  • Shows neuroprotective and immunomodulatory effects
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS