Learn More
Medchemexpress LLC Imipramine-d6 | 65100-45-0 | 500 UG
SDP
Catalog No. 5000411473
Encompass_Preferred
Imipramine-d6 is a deuterium-labeled Imipramine hydrochloride. Imipramine is an orally active tertiary amine tricyclic antidepressant, which acts as a Fascin1 inhibitor with antitumor activities and inhibits serotonin transporter with an IC50 value of 32 nM. It stimulates U-87MG glioma cells autophagy and induces HL-60 cell apoptosis, exhibiting neuroprotective and immunomodulatory effects.
  • Used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • May affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.
  • Fascin1 inhibitor with antitumor activities.
  • Inhibits serotonin transporter (IC50 value of 32 nM).
  • Stimulates U-87MG glioma cells autophagy.
  • Induces HL-60 cell apoptosis.
  • Shows neuroprotective effects.
  • Exhibits immunomodulatory effects.
Catalog No. 50-004-11473 Supplier Medchemexpress LLC Supplier No. HYB1490AS500UG
Imipramine-d6 is a deuterium-labeled Imipramine hydrochloride. Imipramine is an orally active tertiary amine tricyclic antidepressant, which acts as a Fascin1 inhibitor with antitumor activities and inhibits serotonin transporter with an IC50 value of 32 nM. It stimulates U-87MG glioma cells autophagy and induces HL-60 cell apoptosis, exhibiting neuroprotective and immunomodulatory effects.
  • Used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • May affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.
  • Fascin1 inhibitor with antitumor activities.
  • Inhibits serotonin transporter (IC50 value of 32 nM).
  • Stimulates U-87MG glioma cells autophagy.
  • Induces HL-60 cell apoptosis.
  • Shows neuroprotective effects.
  • Exhibits immunomodulatory effects.