Learn More
Medchemexpress LLC HY-B1490 100mg Medchemexpress, Imipramine (hydrochloride) CAS:113-52-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1490100MG
Encompass_Preferred
Medchemexpress, HY-B1490 100mg Imipramine (hydrochloride) CAS:113-52-0 Imipramine hydrochloride inhibits serotonin transporter with an IC 50 value of 32 nM in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2953