Learn More
Medchemexpress LLC Imipramine hydrochloride | 113-52-0 | 99.52% | 100 MG
SDP
Catalog No. 5000411633
Encompass_Preferred
Imipramine hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is an orally active tertiary amine tricyclic antidepressant and a Fascin1 inhibitor with antitumor activities. Additionally, it inhibits serotonin transporter with an IC50 value of 32 nM. This compound stimulates U-87MG glioma cells autophagy, induces HL-60 cell apoptosis, and exhibits neuroprotective and immunomodulatory effects.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appears as a white to off-white solid.
  • Store under recommended conditions in the Certificate of Analysis.
Catalog No. 50-004-11633 Supplier Medchemexpress LLC Supplier No. HYB1490R100MG
Imipramine hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is an orally active tertiary amine tricyclic antidepressant and a Fascin1 inhibitor with antitumor activities. Additionally, it inhibits serotonin transporter with an IC50 value of 32 nM. This compound stimulates U-87MG glioma cells autophagy, induces HL-60 cell apoptosis, and exhibits neuroprotective and immunomodulatory effects.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appears as a white to off-white solid.
  • Store under recommended conditions in the Certificate of Analysis.