Learn More
Medchemexpress LLC Imipramine hydrochloride | 113-52-0 | MFCD00012669 | 99.85% | 1 ML
SDP
Catalog No. 5000411377
Encompass_Preferred
Imipramine hydrochloride is an orally active tertiary amine tricyclic antidepressant. It acts as a Fascin1 inhibitor with antitumor activities and also inhibits the serotonin transporter with an IC50 value of 32 nM. This compound stimulates autophagy in U-87MG glioma cells and induces apoptosis in HL-60 cells. Additionally, it exhibits neuroprotective and immunomodulatory effects.
  • Orally active tertiary amine tricyclic antidepressant.
  • Fascin1 inhibitor with antitumor activities.
  • Inhibits serotonin transporter (IC50 value of 32 nM).
  • Stimulates U-87MG glioma cells autophagy.
  • Induces HL-60 cell apoptosis.
  • Shows neuroprotective effects.
  • Exhibits immunomodulatory effects.
Catalog No. 50-004-11377 Supplier Medchemexpress LLC Supplier No. HYB149010MM/1ML
Imipramine hydrochloride is an orally active tertiary amine tricyclic antidepressant. It acts as a Fascin1 inhibitor with antitumor activities and also inhibits the serotonin transporter with an IC50 value of 32 nM. This compound stimulates autophagy in U-87MG glioma cells and induces apoptosis in HL-60 cells. Additionally, it exhibits neuroprotective and immunomodulatory effects.
  • Orally active tertiary amine tricyclic antidepressant.
  • Fascin1 inhibitor with antitumor activities.
  • Inhibits serotonin transporter (IC50 value of 32 nM).
  • Stimulates U-87MG glioma cells autophagy.
  • Induces HL-60 cell apoptosis.
  • Shows neuroprotective effects.
  • Exhibits immunomodulatory effects.