Learn More
Enzo Life Sciences Imipramine N-Oxide (25mg). CAS: 6829-98-7
SDP
Catalog No. 502011050
Encompass_Preferred
Imipramine metabolite produced via flavin-containing monooxygenase-mediated oxidation. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (30 mg/ml), ethanol (20 mg/ml) or water (2 mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-1050 Supplier Enzo Life Sciences Supplier No. BMLJ1160025
Imipramine metabolite produced via flavin-containing monooxygenase-mediated oxidation. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (30 mg/ml), ethanol (20 mg/ml) or water (2 mg/ml warm). Long Term Storage: Ambient.