Learn More
Medchemexpress LLC HY-B0180 500mg , Imiquimod R 837 CAS:99011-02-6 Purity:98%
SDP
Catalog No. 501492470
Encompass_Preferred
Medchemexpress, HY-B0180 500mg Imiquimod R 837 CAS:99011-02-6 Formula:C14H16N4 Purity:98% Imiquimod is an immune response modifier that acts as a toll-like receptor 7 agonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2470 Supplier Medchemexpress LLC Supplier No. HYB0180500MG
Medchemexpress, HY-B0180 500mg Imiquimod R 837 CAS:99011-02-6 Formula:C14H16N4 Purity:98% Imiquimod is an immune response modifier that acts as a toll-like receptor 7 agonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.