Learn More
Enzo Life Sciences Imiquimod (250mg). CAS: 99011-02-6
SDP
Catalog No. 502009116
Encompass_Preferred
Topical immune response modifier that inhibits angiogenesis. Up-regulates IL-18 and down-regulates MMP-9 through recognition of Toll-like receptor 7 (TLR7) and subsequent activation of MyD88-dependent pathway. Alternative name: 1-(2-Methylpropyl)-1H-imidazo[4,5-c]quinolin-4-amine, R837. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (3mg/ml) or dimethyl formamide; slightly soluble in water (warm to 37°C, vortex); insoluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9116 Supplier Enzo Life Sciences Supplier No. ALX420039M250
Add to Cart
Add to Cart
Topical immune response modifier that inhibits angiogenesis. Up-regulates IL-18 and down-regulates MMP-9 through recognition of Toll-like receptor 7 (TLR7) and subsequent activation of MyD88-dependent pathway. Alternative name: 1-(2-Methylpropyl)-1H-imidazo[4,5-c]quinolin-4-amine, R837. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (3mg/ml) or dimethyl formamide; slightly soluble in water (warm to 37°C, vortex); insoluble in 100% ethanol. Long Term Storage: -20°C.