Learn More
TARGETMOL CHEMICALS INC Imiquimod 500MG
Supplier: TARGETMOL CHEMICALS INC T0134500MG
Also available in 1 g, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Imiquimod (R 837) is an immune response modifier that acts as a toll-like receptor 7 agonist. Purity 99.66%