Learn More
Apexbio Technology LLC Imiquimod 99011-02-6 100mg
SDP
Supplier:  Apexbio Technology LLC A5161100
Encompass_Preferred
Imiquimod (CAS 99011-02-6) is a small-molecule agonist targeting Toll-like receptor 7 (TLR7) It is designed to activate TLR7 thereby modulating innate immune responses Imiquimod exerts its biological activity primarily through induction of cytokines such as interferon-gamma and interleukin-12 while suppressing IL-4 and IL-5 thus affecting cellular immunity In in vitro studies imiquimod demonstrates TLR7 activation with an EC50 of approximately 2 1 M and exhibits an anti-proliferative effect mediated by opioid growth factor (OGF) with an IC50 of 2 M Based on these pharmacological properties imiquimod holds research potential in cancer inflammatory diseases and dermatologic conditions including basal cell carcinoma superficial squamous cell carcinoma actinic keratosis and external genital warts
Catalog No. 50-000-92844