Learn More
Medchemexpress LLC HY-B0180A 200mg Medchemexpress, Imiquimod (hydrochloride) CAS:99011-78-6 Purity:>98%
SDP
Catalog No. 502082398
Encompass_Preferred
Medchemexpress, HY-B0180A 200mg Imiquimod (hydrochloride) CAS:99011-78-6 Imiquimod hydrochloride (R 837 hydrochloride) is a selective toll like receptor 7 (TLR7) agonist acting as an immune response modifier. Imiquimod hydrochloride exhibits antiviral and antitumor effects in vivo. Imiquimod hydrochloride can be used for the research of external genital, perianal warts, cancer and COVID 19. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2398 Supplier Medchemexpress LLC Supplier No. HYB0180A200MG
Medchemexpress, HY-B0180A 200mg Imiquimod (hydrochloride) CAS:99011-78-6 Imiquimod hydrochloride (R 837 hydrochloride) is a selective toll like receptor 7 (TLR7) agonist acting as an immune response modifier. Imiquimod hydrochloride exhibits antiviral and antitumor effects in vivo. Imiquimod hydrochloride can be used for the research of external genital, perianal warts, cancer and COVID 19. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Product Title
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.