Learn More
Medchemexpress LLC Imiquimod (hydrochloride) | 99011-78-6 | 10 MM 1 ML
SDP
Catalog No. 5000406627
Encompass_Preferred
Imiquimod hydrochloride, also known as R 837 hydrochloride, is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. This compound exhibits antiviral and antitumor effects in vivo, making it valuable for various research applications.
  • Used for research of external genital, perianal warts, cancer, and COVID-19
  • Can induce psoriasis models
  • Exhibits antiviral effects in vivo
  • Exhibits antitumor effects in vivo
  • Acts as a selective toll-like receptor 7 (TLR7) agonist
  • Functions as an immune response modifier
Catalog No. 50-004-06627 Supplier Medchemexpress LLC Supplier No. HYB0180A10MM/1ML
Imiquimod hydrochloride, also known as R 837 hydrochloride, is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. This compound exhibits antiviral and antitumor effects in vivo, making it valuable for various research applications.
  • Used for research of external genital, perianal warts, cancer, and COVID-19
  • Can induce psoriasis models
  • Exhibits antiviral effects in vivo
  • Exhibits antitumor effects in vivo
  • Acts as a selective toll-like receptor 7 (TLR7) agonist
  • Functions as an immune response modifier