Learn More
Medchemexpress LLC Imiquimod hydrochloride | 99011-78-6 | 99.8% | 100 MG  DFS Item
Catalog No. 5000405934
Imiquimod hydrochloride (R 837 hydrochloride) is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It exhibits antiviral and antitumor effects in vivo and can be used for the research of external genital, perianal warts, cancer, and COVID-19.
  • Exhibits antiviral effects in vivo
  • Exhibits antitumor effects in vivo
  • Selective TLR7 agonist
  • Used in research for external genital, perianal warts, cancer, and COVID-19
Catalog No. 50-004-05934 Supplier Medchemexpress LLC Supplier No. HYB0180A100MG
Imiquimod hydrochloride (R 837 hydrochloride) is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It exhibits antiviral and antitumor effects in vivo and can be used for the research of external genital, perianal warts, cancer, and COVID-19.
  • Exhibits antiviral effects in vivo
  • Exhibits antitumor effects in vivo
  • Selective TLR7 agonist
  • Used in research for external genital, perianal warts, cancer, and COVID-19