Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imiquimod maleate | 896106-16-4 | 99.9% | 100 MG
SDP

Catalog No. 5000406177
Encompass_Preferred

Imiquimod maleate is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in vivo and is used in research for external genital and perianal warts, cancer, and COVID-19. It appears as a white to off-white solid.

  • Molecular weight: 356.38
  • Formula: C18H20N4O4
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (25 mg/mL) and H2O (2 mg/mL)
  • Targets TLR7 and HSV-1
  • Used to induce psoriasis models

Catalog No. 50-004-06177 Supplier Medchemexpress LLC Supplier No. HYB0180B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imiquimod maleate is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in vivo and is used in research for external genital and perianal warts, cancer, and COVID-19. It appears as a white to off-white solid.

  • Molecular weight: 356.38
  • Formula: C18H20N4O4
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (25 mg/mL) and H2O (2 mg/mL)
  • Targets TLR7 and HSV-1
  • Used to induce psoriasis models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.