Learn More
Medchemexpress LLC Imiquimod maleate | 896106-16-4 | 99.9% | 200 MG
SDP
Catalog No. 5000405935
Encompass_Preferred
Imiquimod maleate is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It exhibits antiviral and antitumor effects in vivo. This product is used for research related to external genital, perianal warts, cancer, and COVID-19.
  • Immune response modifier
  • Selective toll-like receptor 7 (TLR7) agonist
  • Exhibits antiviral and antitumor effects in vivo
  • Used for research of external genital, perianal warts, cancer, and COVID-19
  • Can be used to induce psoriasis models
Catalog No. 50-004-05935 Supplier Medchemexpress LLC Supplier No. HYB0180B200MG
Imiquimod maleate is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It exhibits antiviral and antitumor effects in vivo. This product is used for research related to external genital, perianal warts, cancer, and COVID-19.
  • Immune response modifier
  • Selective toll-like receptor 7 (TLR7) agonist
  • Exhibits antiviral and antitumor effects in vivo
  • Used for research of external genital, perianal warts, cancer, and COVID-19
  • Can be used to induce psoriasis models