Learn More
Medchemexpress LLC Imiquimod (maleate) | 896106-16-4 | 99.9% | 500 MG
SDP
Catalog No. 5000406628
Encompass_Preferred
Imiquimod maleate (R 837 maleate) is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in vivo. This product is an analytical standard intended for research and analytical applications, and can be used for research of external genital, perianal warts, cancer, and COVID-19, and to induce psoriasis models.
  • Selective toll-like receptor 7 (TLR7) agonist
  • Exhibits antiviral and antitumor effects in vivo
  • Suitable for research of external genital, perianal warts, cancer, and COVID-19
  • Can be used to induce psoriasis models
  • Targets TLR7 and HSV-1
Catalog No. 50-004-06628 Supplier Medchemexpress LLC Supplier No. HYB0180B500MG
Imiquimod maleate (R 837 maleate) is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in vivo. This product is an analytical standard intended for research and analytical applications, and can be used for research of external genital, perianal warts, cancer, and COVID-19, and to induce psoriasis models.
  • Selective toll-like receptor 7 (TLR7) agonist
  • Exhibits antiviral and antitumor effects in vivo
  • Suitable for research of external genital, perianal warts, cancer, and COVID-19
  • Can be used to induce psoriasis models
  • Targets TLR7 and HSV-1