Learn More
Invivogen Imiquimod (R837) TLR7 Agonist, 10 mg Sterile Water Included, Purity ≥95%
SDP
Catalog No. NC2684138
Encompass
Imiquimod (R837) is a potent TLR7 agonist designed for precise immune system modulation, inducing pro-inflammatory cytokines and enhancing antiviral and antitumor responses. Each set includes 10 mg of purified compound alongside sterile endotoxin-free water. With high purity and rigorous contamination testing, it serves as an effective adjuvant, differentiating itself by targeting specific immune pathways in both human and mouse models.
Catalog No. NC2684138 Supplier Invivogen Supplier No. TLRLIMQ10
Imiquimod (R837) is a potent TLR7 agonist designed for precise immune system modulation, inducing pro-inflammatory cytokines and enhancing antiviral and antitumor responses. Each set includes 10 mg of purified compound alongside sterile endotoxin-free water. With high purity and rigorous contamination testing, it serves as an effective adjuvant, differentiating itself by targeting specific immune pathways in both human and mouse models.