Learn More
Medchemexpress LLC Imirestat | 89391-50-4 | 99.6% | 286.24 | 100 MG
SDP
Catalog No. 5000392445
Encompass_Preferred
Imirestat (AL 1576) is an aldose reductase inhibitor used for the treatment of diabetes. For research use only, MedChemExpress does not sell to patients.
  • Aldose reductase inhibitor
  • Used for the treatment of diabetes
  • Purity: 99.59%
  • Improves nerve conduction velocity
Catalog No. 50-003-92445 Supplier Medchemexpress LLC Supplier No. HY16255100MG
Imirestat (AL 1576) is an aldose reductase inhibitor used for the treatment of diabetes. For research use only, MedChemExpress does not sell to patients.
  • Aldose reductase inhibitor
  • Used for the treatment of diabetes
  • Purity: 99.59%
  • Improves nerve conduction velocity