Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imirestat | 89391-50-4 | 99.6% | 286.24 | 50 MG
SDP

Catalog No. 5000391338
Encompass_Preferred

Imirestat, also known as AL 1576, is an aldose reductase inhibitor primarily used for the treatment of diabetes. This compound is intended for research use only.

  • High purity: 99.6%
  • Molecular weight: 286.24
  • Chemical formula: C15H8F2N2O2
  • Appearance: White to off-white solid
  • Optimal storage for powder: -20°C for 3 years, 4°C for 2 years
  • Optimal storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO

Catalog No. 50-003-91338 Supplier Medchemexpress LLC Supplier No. HY1625550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imirestat, also known as AL 1576, is an aldose reductase inhibitor primarily used for the treatment of diabetes. This compound is intended for research use only.

  • High purity: 99.6%
  • Molecular weight: 286.24
  • Chemical formula: C15H8F2N2O2
  • Appearance: White to off-white solid
  • Optimal storage for powder: -20°C for 3 years, 4°C for 2 years
  • Optimal storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.