Learn More
Thermo Scientific™ Imject™ Maleimide-Activated BSA
Catalog No. p-4527378 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77115 10 mg
PI77116 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI77115 Supplier Thermo Scientific™ Supplier No. 77115
Add to Cart
Edge
Add to Cart
Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.
Thermo Scientific™ Imject Maleimide-Activated BSA is sulfhydryl-reactive bovine serum albumin for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Purified, carrier-grade BSA is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Several antigen molecules can be conjugated to each BSA molecule to increase their immunogenicity and likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest. Select from among several package sizes of activated BSA protein, or choose the complete conjugation kit that includes optimized buffers and trouble-free spin desalting columns.
Highlights:
  • Convenient packages – choose stabilized, lyophilized Maleimide-Activated BSA or complete immunogen conjugation kit
  • Optimized activation – specifications require that each lot is activated with 15 to 20 moles of sulhydryl-reactive maleimide per mole of BSA, ensuring consistently high levels of conjugation
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – pure BSA (Fraction V) is activated using high-quality Thermo Scientific Sulfo-SMCC crosslinker and formulated to ensure high stability and consistent performance
  • Immunogenic and soluble – moderately immunogenic and highly soluble, BSA is useful for preparation of the primary immunogen or as an irrelevant hapten-carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
Includes:
Kit contains stabilized, lyophilized Maleimide-Activated BSA (in PBS with stabilizer), buffers and desalting columns to easily prepare ready-to-use immunogens for immunization
TRUSTED_SUSTAINABILITY
Specifications
Product Line Imject
Product Type Imject Maleimide Activated Bovine Serum Albumin
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Quantity 10 mg
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.