Learn More
Description
Thermo Scientific™ Imject Maleimide-Activated mcKLH is sulfhydryl-reactive immunogenic mariculture keyhole limpet hemocyanin (KLH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Highlights:
Convenient packages – choose stabilized, lyophilized Maleimide-Activated mcKLH or a complete immunogen conjugation kit
Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of KLH, ensuring consistently high levels of conjugation
Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein – KLH is isolated from the mollusk Megathura crenulata (keyhole limpet); mcKLH is a high-quality form of KLH that is harvested from limpets grown in mariculture
Highly immunogenic – KLH has a high molecular mass (0.5 to 13 million daltons; aggregates of 350 and 390kDa subunits) and elicits a stronger immune response than BSA or ovalbumin
Excellent solubility – our preparations of mariculture KLH are more soluble than typical forms and we also offer a PEGylated form for even greater hydrophilicity and solubility
Includes:
Kit contains stabilized, lyophilized Maleimide-Activated mcKLH (in PBS with stabilizer), buffers and desalting columns to easily prepare ready-to-use immunogens for immunization
Specifications
Specifications
| Product Line | Imject |
| Quantity | 5 x 2 mg |
| Content And Storage | Sufficient For: 5 conjugation reactions and complete immunogen preparations, each with up to 2 mg sulfhydryl-containing peptide • Imject Maleimide-Activated mcKLH, 5 x 2 mg • Imject Maleimide Conjugation Buffer, 30 mL • Imject Purification Buffer Salts, 5 x 0.84 g • Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns Store the Maleimide-Activated blue Carrier Protein at -20°C. Store remaining components at 4°C. |
| Description | Imject Maleimide Activated mcKLH Spin Kit |
| Format | Kit |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.