Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Imject™ Ovalbumin (in PBS)

Catalog No. PI77120 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77120 5 x 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI77120 Supplier Thermo Scientific™ Supplier No. 77120
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepare hapten-carrier conjugates to generate antibodies using this conveniently formulated, purified ovalbumin (OVA) protein.

Thermo Scientific™ Imject™ Ovalbumin Carrier Protein is purified ovalbumin protein conveniently formulated for preparation of hapten-carrier conjugates.

Because ovalbumin is only weakly immunogenic, its primary use in antibody production workflows is as a secondary (independent) carrier protein to make antigens more amenable to antibody screening procedures. For example, peptide antigens are more easily coated in polystyrene microplates when conjugated to ovalbumin, thereby facilitating the screening of antisera that were produced using immunogen conjugates with KLH or other more immunogenic carrier protein.

Highlights:

  • Purified from chicken egg whites and lyophilized in phosphate buffered saline
  • Single polypeptide protein with molecular mass of 45,000 daltons
  • Contains 20 lysine residues, most of which have primary amines that are capable of reacting with a conjugation reagent
  • Contains 14 aspartic acid and 33 glutamic acid residues whose carboxyl groups account for the net negative charge (pI 4.63) of ovalbumin
  • Not as immunogenic as keyhole limpet hemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent
  • Useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
TRUSTED_SUSTAINABILITY

Specifications

Product Line Imject
Quantity 5 x 20 mg
Content And Storage Supplied: Carrier proteins are supplied lyophilized. When reconstituted with 2mL of ultrapure water the buffer compositions for BSA and mcKLH is 0.1M sodium phosphate, 0.15M NaCl; pH 7.2 and for OVA is 0.01M sodium phosphate, pH 7.2. Proteins contain proprietary stabilizers.

Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.