Learn More
Creative Biomart Recombinant Zika Ectodomain Antigen
Catalog No. 502085078
Encompass_Preferred
Recombinant Zika Ectodomain is a glycosylated protein, produced using baculovirus vectors in insect cells and its Mw is approximately 45 kDa.The Zika Ectodomain is a recombinant protein of the ectodom
Catalog No. 50-208-5078 Supplier Creative Biomart Supplier No. VAC0238
Recombinant Zika Ectodomain is a glycosylated protein, produced using baculovirus vectors in insect cells and its Mw is approximately 45 kDa.The Zika Ectodomain is a recombinant protein of the ectodom