Learn More
Andwin Scientific Yamato Immersion Cooler Device, Stainless Steel Flexible Tube Cooling Coil, 500mm length, 115V/220V
SDP
Catalog No. 501898172
Encompass_Preferred
Easy to use benchtop immersion cooler with stainless steel flexible tube cooling coil used in combination with water bath. Cools down water bath immediately by dipping cooling coil into fluid. Stainless steel flexible cooling coil is high corrosion resistant. Improved safety by adopting overcurrent circuit breaker. Comes with handles for better usability. Temperature range from -20 ~ +35°C.
Catalog No. 50-189-8172 Supplier Andwin Scientific Supplier No. BE201F115V
Easy to use benchtop immersion cooler with stainless steel flexible tube cooling coil used in combination with water bath. Cools down water bath immediately by dipping cooling coil into fluid. Stainless steel flexible cooling coil is high corrosion resistant. Improved safety by adopting overcurrent circuit breaker. Comes with handles for better usability. Temperature range from -20 ~ +35°C.