Learn More
Volu Sol Immersion Oil, Type B (High Viscosity), 4 oz / 120 mL, Volu-Sol™
SDP
Catalog No. 50420878 Shop All Volu Sol Products
Encompass_Preferred
Types A and B are essentially interchangeable. Oils are miscible with each other, can be mixed for intermediate viscosities. Type A, at 150 centistokes, reduces most tendencies to trap air, being most beneficial to researchers. Air bubbles can cause image degradation. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application
Catalog No. 50-420-878 Supplier Volu Sol Supplier No. 164844
Types A and B are essentially interchangeable. Oils are miscible with each other, can be mixed for intermediate viscosities. Type A, at 150 centistokes, reduces most tendencies to trap air, being most beneficial to researchers. Air bubbles can cause image degradation. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application