Learn More
Ace Glass, Inc. Immersion well, complete, 220mm
SDP
Catalog No. 50979575
Encompass
220mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.
Catalog No. 50-979-575 Supplier Ace Glass, Inc. Supplier No. 785842
220mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.