Learn More
Ace Glass, Inc. Immersion well, complete outer chamber, 290mm
SDP
Catalog No. 50975761
Encompass
290mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.
Catalog No. 50-975-761 Supplier Ace Glass, Inc. Supplier No. 785814
290mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.